Order Entry
Germany
ContactUsLinkComponent
Succinate, 0,2 M buffer solution, pH 6,5
Succinate, 0,2 M buffer solution, pH 6,5
Certificates
Succinate, 0,2 M buffer solution, pH 6,5
Certificates

Specifications

  • CAS No.:
    150-90-3
  • MDL No.:
    MFCD00002789
  • Storage temperature:
    2°C to 8°C
  • Physical form:
    Liquid
  • Odour/scent:
    Odourless
  • Appearance:
    Colourless

Specifications

Related Information