Order Entry
Germany
ContactUsLinkComponent
MOPSO, 0,2 M buffer soln., pH 7,0
MOPSO, 0,2 M buffer soln., pH 7,0
Certificates
MOPSO, 0,2 M buffer soln., pH 7,0
Certificates

Specifications

  • CAS No.:
    79803-73-9
  • MDL No.:
    MFCD00070007
  • Physical form:
    Liquid

Specifications

Related Information