Order Entry
Germany
ContactUsLinkComponent
Phosphate, 0,2 M buffer solution, pH 7,5
Phosphate, 0,2 M buffer solution, pH 7,5
Certificates
Phosphate, 0,2 M buffer solution, pH 7,5
Certificates

Specifications

  • CAS No.:
    7558-80-7
  • MDL No.:
    MFCD00003527
  • Physical form:
    Liquid

Specifications

Related Information