
(+/-)-Octopamine hydrochloride 99%
Catalog # J61281.09
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:10 g
Specifications
About this item
CAS Number:770-05-8
:
Formula:C₈H₁₂ClNO₂
MDL Number:MFCD00012881
Merck Index:14,6759
Melting Pt:170 °C (dec.)
:189.639 g/mol
EINECS:212-216-4
:GHS07
Storage Temperature:Ambient