

Fluoxetine hydrochloride 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:56296-78-7
:59333-67-4
Formula:C₁₇H₁₈F₃NO·HCl
MDL Number:MFCD00214288
:345.79 g/mol
Merck Index:14,4185
Melting Pt:158…159 °C
EINECS:260-101-2
:GHS05
:GHS07
Storage Temperature:Ambient