Order Entry
Germany
Orders LinkContactUsLinkComponent
Phosphate, 0,5M buffer solution, pH 8,5

Phosphate, 0,5M buffer solution, pH 8,5

Phosphate, 0,5M buffer solution, pH 8,5
Certificates
Phosphate, 0,5M buffer solution, pH 8,5

2 Options Available Below

Product Details & Documents

CAS No.: 7558-80-7
Physical Form: Liquid
MDL No.: MFCD00003527

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • J61086.AK
  • J61086.AP
  • Physical form:
  • Liquid
  • Liquid
  • CAS No.:
  • 7558-80-7
  • 7558-80-7
  • MDL No.:
  • MFCD00003527
  • MFCD00003527

Specifications

Product Family Options

Product Information

  • Concentration
    pH value
    Pk
    Availability
    Price
  • J61086.AK
    Phosphate buffer soln
    Phosphate buffer soln
    0,5 M
    8,5
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 68,20 €
    68,20 €
    1 * 250 mL
       68,20 €

    Specifications:
    More
    Specifications
    Cat. No.
    J61086.AK
    Physical form
    Liquid
    CAS No.
    7558-80-7
    MDL No.
    MFCD00003527
  • J61086.AP
    Phosphate buffer soln
    Phosphate buffer soln
    0,5 M
    8,5
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 92,40 €
    92,40 €
    1 * 500 mL
       92,40 €

    Specifications:
    More
    Specifications
    Cat. No.
    J61086.AP
    Physical form
    Liquid
    CAS No.
    7558-80-7
    MDL No.
    MFCD00003527
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.