Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Vinpocetine
Vinpocetine 98%
Vinpocetine 98%
Vinpocetine 98%
J61000.MC
:
Thermo Fisher Scientific
:
Vinpocetine 98%
J61000.MC
:
Thermo Fisher Scientific
:
Pk:
100 mg
:
:
350.46 g/mol
:
14,9991
:
147…149 °C
:
256-028-0
:
GHS07
:
Ambient
:
42971-09-5
:
C₂₂H₂₆N₂O₂
:
MFCD00211233