PIPES lysis buffer with NP-40 (2X)
Catalog # J60568.AK
Lieferant: Thermo Fisher Scientific
CAS-Nummer:
Spezifikationen
- Pk:250 mL
- Description:PIPES lysis buffer with NP-40 (2X)
- Physical form:Viscous liquid
- Odour/scent:Odourless
- Appearance:Colourless
Spezifikationen
Über diesen Artikel
Physical Form: Viscous liquid
Appearance: Colourless
Odour: Odourless