Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Amiodarone hydrochloride
Amiodarone hydrochloride
Amiodarone hydrochloride
Amiodarone hydrochloride
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Amiodarone hydrochloride
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
19774-82-4
:
Formula:
C₂₅H₂₉I₂NO₃
MDL Number:
MFCD00069204
:
681.78 g/mol
Merck Index:
14,482
Melting Pt:
156…163 °C
EINECS:
243-293-2
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAJ60456
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation