EDTA (ethylenediamine tetraacetic acid) 0.5 M in aqueous solution, autoclaved pH 8.0
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:60-00-4
:
Formula:C₁₀H₁₆N₂O₈
MDL Number:MFCD00003541
:292.25 g/mol
Storage Temperature:Ambient