
3-Fluoro-4-hydroxy-5-methoxybenzaldehyde 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:79418-78-3
:
Formula:C₈H₇FO₃
MDL Number:MFCD02683560
:170.139 g/mol
Boiling Pt:114…118 °C
:GHS07
Storage Temperature:Ambient