Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 5-bromo-2-chloronicotinate
Methyl-5-bromo-2-chloronicotinate 98%
Methyl-5-bromo-2-chloronicotinate 98%
:
Thermo Fisher Scientific
Methyl-5-bromo-2-chloronicotinate 98%
:
Thermo Fisher Scientific
:
78686-79-0
:
:
C₇H₅BrClNO₂
:
MFCD03844848
:
250.48 g/mol
:
48…53 °C
:
>110 °C (230 °F)
:
GHS07
:
Ambient
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years