1-Bromo-4-fluorobutane 97%
Catalog # H64317.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:155.01 g/mol
Boiling Pt:134…135 °C
Density:1.444
Flash Pt:45 °C (113 °F)
UN:1993
ADR:3,III
:GHS02
:GHS07
Storage Temperature:Ambient
CAS Number:462-72-6
Formula:BrH₂C(CH₂)₂CH₂F
MDL Number:MFCD00209551