Order Entry
Germany
ContactUsLinkComponent
1-tert-Butyl-4-ethyl-3-oxopiperidine-1,4-dicarboxylate 97%
1-tert-Butyl-4-ethyl-3-oxopiperidine-1,4-dicarboxylate 97%
Catalog # H64102.06
CAS Number:  
1-tert-Butyl-4-ethyl-3-oxopiperidine-1,4-dicarboxylate 97%
Catalog # H64102.06
CAS Number:  

Specifications

  • Pk:
    5 g

Specifications

About this item

CAS Number:71233-25-5
 :
Formula:C₁₃H₂₁NO₅
 :271.31 g/mol
 :GHS07
Storage Temperature:Ambient
MDL Number:MFCD09878815