

2-(Trifluoromethyl)pyridine-5-carboxaldehyde 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:386704-12-7
:
Formula:C₇H₄F₃NO
MDL Number:MFCD01862647
:175.11 g/mol
Melting Pt:52…56 °C
Flash Pt:93 °C (199 °F)
ADR:6.1,III
:GHS06
:GHS07