Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
HMDS (1,1,1,3,3,3-Hexamethyldisilazane)
HMDS (1,1,1,3,3,3-hexamethyldisilazane) ≥98%
HMDS (1,1,1,3,3,3-hexamethyldisilazane) ≥98%
:
Thermo Fisher Scientific
HMDS (1,1,1,3,3,3-hexamethyldisilazane) ≥98%
:
Thermo Fisher Scientific
:
999-97-3
:
:
(CH₃)₃SiNHSi(CH₃)₃
:
MFCD00008259
:
161.39 g/mol
:
14,4689
:
126 °C
:
–78 °C
:
0.774
:
8 °C (46 °F)
:
Ambient
:
213-668-5
:
3286
:
3,II
:
01-2119438176-38
:
GHS02
:
GHS06
:
GHS05
msds_ALFAA15139
Thermo Scientific Acroseal 25 Years