
Methyl (R)-3-[bis(4-methoxyphenyl)methyl]-2,2-dioxo[1,2,3]oxathiazolidine-4-carboxylate 97%
Catalog # H63889.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:5 g
Specifications
About this item
CAS Number:1212136-00-9
:
Formula:C₁₉H₂₁NO₇S
:407.437 g/mol
Melting Pt:100…107 °C
:GHS07
MDL Number:MFCD09864994