Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Iodo-5-trifluoromethylpyridine
2-Iodo-5-trifluoromethylpyridine 96%
2-Iodo-5-trifluoromethylpyridine 96%
Irritant
2-Iodo-5-trifluoromethylpyridine 96%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
2-Iodo-5-trifluoromethylpyridine 96%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
100366-75-4
:
Formula:
C₆H₃F₃IN
MDL Number:
MFCD08741351
:
272.99 g/mol
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation