Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
trans-N-Fmoc-4-tert-butoxy-L-proline
trans-N-Fmoc-4-tert-butoxy-L-proline 98%
trans-N-Fmoc-4-tert-butoxy-L-proline 98%
trans-N-Fmoc-4-tert-butoxy-L-proline 98%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
trans-N-Fmoc-4-tert-butoxy-L-proline 98%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
122996-47-8
:
Formula:
C₂₄H₂₇NO₅
MDL Number:
MFCD00151930
:
409.482 g/mol
Related Information
Documentation
Certificates
Search by Lot Number
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation