Di-tert-butyl L-aspartate hydrochloride 98%
Catalog # H63716.14
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:25 g
Specifications
About this item
CAS Number:1791-13-5
:
Formula:C₁₂H₂₃NO₄
MDL Number:MFCD00034851
:281.78 g/mol