Order Entry
Germany
ContactUsLinkComponent
7-Chloro-1,2,3,4-tetrahydroisoquinoline 95%
Irritant
7-Chloro-1,2,3,4-tetrahydroisoquinoline 95%
Catalog # H63693.03
CAS Number:  
Irritant
7-Chloro-1,2,3,4-tetrahydroisoquinoline 95%
Catalog # H63693.03
CAS Number:  

Specifications

  • Pk:
    1 g

Specifications

About this item

 :
 :167.636 g/mol
 :GHS07
CAS Number:82771-60-6
Formula:C₉H₁₀ClN
MDL Number:MFCD05861543