Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
H-d-glu-otbu
H-d-glu-otbu 98%
H-d-glu-otbu 98%
:
Thermo Fisher Scientific
H-d-glu-otbu 98%
:
Thermo Fisher Scientific
:
25456-76-2
:
:
C₉H₁₇NO₄
:
MFCD09842819
:
203.238 g/mol
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years