0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Boc-glycinamide
Boc-glycinamide 95%
Boc-glycinamide 95%
:
Thermo Fisher Scientific
Boc-glycinamide 95%
:
Thermo Fisher Scientific
:
35150-09-5
:
:
C₇H₁₄N₂O₃
:
MFCD08275106
:
174.2 g/mol
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years