Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Boc-Phe(4-CN)-OH
Boc-Phe(4-CN)-OH 95%
Boc-Phe(4-CN)-OH 95%
Boc-Phe(4-CN)-OH 95%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Boc-Phe(4-CN)-OH 95%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
131724-45-3
:
Formula:
C₁₅H₁₈N₂O₄
MDL Number:
MFCD00672526
Melting Pt:
156…160 °C
ADR:
6.1,III
:
290.319 g/mol
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation