1-Boc-3-(ethylamino)azetidine 95%
Catalog # H57941.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
CAS Number:454703-23-2
:
Formula:C₁₀H₂₀N₂O₂
MDL Number:MFCD11520619
:200.28 g/mol
:GHS07
Storage Temperature:Ambient