Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
D-beta-Proline ethyl ester hydrochloride
D-β-Proline ethyl ester hydrochloride 97%
D-β-Proline ethyl ester hydrochloride 97%
H57789.03
:
Thermo Fisher Scientific
:
D-β-Proline ethyl ester hydrochloride 97%
H57789.03
:
Thermo Fisher Scientific
:
Pk:
1 g
Seq:
3047
:
:
C₇H₁₃NO₂·HCl
:
MFCD18071037
:
179.65 g/mol
:
GHS07
:
Refrigerator