Order Entry
Germany
ContactUsLinkComponent
 
Irritant
(S)-1-Boc-4-oxo-piperidine-2-carboxylic acid methyl ester 95%
Catalog # H57213.03
CAS Number:  
Irritant
(S)-1-Boc-4-oxo-piperidine-2-carboxylic acid methyl ester 95%
Catalog # H57213.03
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

CAS Number:180854-44-8
 :
Formula:C₁₃H₂₁NO₅
 :271.32 g/mol
Density:1.079
Flash Pt:>110 °C (230 °F)
 :GHS07
MDL Number:MFCD15071881