Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl-4-hydroxy-3-iodobenzoate
Methyl-4-hydroxy-3-iodobenzoate 98%
Methyl-4-hydroxy-3-iodobenzoate 98%
H56877.06
:
Thermo Fisher Scientific
:
Methyl-4-hydroxy-3-iodobenzoate 98%
H56877.06
:
Thermo Fisher Scientific
:
Pk:
5 g
:
15126-06-4
:
:
C₈H₇IO₃
:
278.04 g/mol
:
155…159 °C
:
GHS05
:
GHS07
:
Ambient
:
MFCD05664378