Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Boc-4-(6-nitro-3-pyridyl)piperazine
1-Boc-4-(6-nitro-3-pyridyl)piperazine 97%
1-Boc-4-(6-nitro-3-pyridyl)piperazine 97%
1-Boc-4-(6-nitro-3-pyridyl)piperazine 97%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
1-Boc-4-(6-nitro-3-pyridyl)piperazine 97%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
571189-16-7
:
Formula:
C₁₄H₂₀N₄O₄
MDL Number:
MFCD11849291
:
308.33 g/mol
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAH54465
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation