Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Boc-4'-(Bromomethyl)biphenyl
2-Boc-4'-(Bromomethyl)biphenyl 97%
2-Boc-4'-(Bromomethyl)biphenyl 97%
2-Boc-4'-(Bromomethyl)biphenyl 97%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
2-Boc-4'-(Bromomethyl)biphenyl 97%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
114772-40-6
:
Formula:
C₁₈H₁₉BrO₂
MDL Number:
MFCD06657561
:
347.24 g/mol
EINECS:
442-850-9
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAH54100
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation