Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
tert-Butyl-4-formylpiperidine-1-carboxylate
tert-Butyl-4-formylpiperidine-1-carboxylate 97%
tert-Butyl-4-formylpiperidine-1-carboxylate 97%
tert-Butyl-4-formylpiperidine-1-carboxylate 97%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
tert-Butyl-4-formylpiperidine-1-carboxylate 97%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
137076-22-3
:
Formula:
C₁₁H₁₉NO₃
MDL Number:
MFCD02179019
:
213.28 g/mol
:
GHS07
Storage Temperature:
Refrigerator
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAH52813
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation