
1-Boc-3-hydroxy-1,2,3,6-tetrahydropyridine 97%
Catalog # H52577.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:199.25 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:224779-27-5
Formula:C₁₀H₁₇NO₃
MDL Number:MFCD04972246