tert-Butylmagnesium chloride 1 M in MeTHF (2-methyltetrahydrofuran)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:677-22-5
:
Formula:C₄H₉ClMg
MDL Number:MFCD00000465
:116.87 g/mol
UN:3399
ADR:4.3,II
:GHS02
:GHS05
Storage Temperature:Ambient