
(R)-(+)-2-Amino-1,1-diphenyl-1-propanol 99%
Catalog # H51083.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
CAS Number:78603-93-7
:
Formula:C₁₅H₁₇NO
MDL Number:MFCD02677967
:227.30 g/mol
Melting Pt:102…105 °C
Storage Temperature:Ambient
ADR:8,III
:GHS05