0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
2-Cyclohexyloxypyridine-5-carboxaldehyde
2-Cyclohexyloxypyridine-5-carboxaldehyde 97%
2-Cyclohexyloxypyridine-5-carboxaldehyde 97%
H50129.03
:
Thermo Fisher Scientific
:
2-Cyclohexyloxypyridine-5-carboxaldehyde 97%
H50129.03
:
Thermo Fisher Scientific
:
Pk:
1 g
Seq:
3047
:
916792-16-0
:
:
C₁₂H₁₅NO₂
:
MFCD08060944
:
205.26 g/mol
:
GHS07
:
Ambient