Order Entry
Germany
ContactUsLinkComponent
2-(Boc-amino)pyridine-3-boronic acid pinacol ester 98%
2-(Boc-amino)pyridine-3-boronic acid pinacol ester 98%
Catalog # H50094.03
CAS Number:  
2-(Boc-amino)pyridine-3-boronic acid pinacol ester 98%
Catalog # H50094.03
CAS Number:  

Specifications

  • Pk:
    1 g

Specifications

About this item

CAS Number:1072944-99-0
 :
Formula:C₁₆H₂₅BN₂O₄
 :320.196 g/mol
 :GHS07
Storage Temperature:Ambient
MDL Number:MFCD08063072