1-(tert-Butoxycarbonyl)pyrrolidine-3-carboxylic acid 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:59378-75-5
:
Formula:C₁₀H₁₇NO₄
MDL Number:MFCD02179181
:215.25 g/mol
Melting Pt:128…137 °C
:GHS07
Storage Temperature:Ambient