Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Registrieren
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Pyrazolo[1,5-a]pyridine-7-carbaldehyde
Pyrazolo[1,5-a]pyridine-7-carbaldehyde 97%
Pyrazolo[1,5-a]pyridine-7-carbaldehyde 97%
Irritant
Pyrazolo[1,5-a]pyridine-7-carbaldehyde 97%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
Pyrazolo[1,5-a]pyridine-7-carbaldehyde 97%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
362661-83-4
:
Formula:
C₈H₆N₂O
MDL Number:
MFCD06739029
:
146.15 g/mol
Melting Pt:
83…86 °C
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAH50074
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation