Order Entry
Germany
ContactUsLinkComponent
Palladium 5% on calcium carbonate lead poisoned A305060-5
Health
Palladium 5% on calcium carbonate lead poisoned A305060-5
Catalog # H36186.06
CAS Number:  
Health
Palladium 5% on calcium carbonate lead poisoned A305060-5
Catalog # H36186.06
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :106.420 g/mol
EINECS:231-115-6
 :GHS08
Storage Temperature:Ambient
CAS Number:7440-05-3
Formula:Pd
MDL Number:MFCD03427452