tert-Butyl-2-(hydroxymethyl)pyrrolidine-1-carboxylate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:170491-63-1
:
Formula:C₁₀H₁₉NO₃
MDL Number:MFCD01456556
:201.3 g/mol
Melting Pt:52…54 °C
:GHS07
Storage Temperature:Ambient