Order Entry
Germany
ContactUsLinkComponent
4-(Aminomethyl)pyrimidine hydrochloride 97%
Irritant
4-(Aminomethyl)pyrimidine hydrochloride 97%
Catalog # H32582.ME
CAS Number:  
Irritant
4-(Aminomethyl)pyrimidine hydrochloride 97%
Catalog # H32582.ME
CAS Number:  

Specifications

  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :145.59 g/mol
Melting Pt:176…178 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:1138011-17-2
Formula:C₅H₇N₃·HCl
MDL Number:MFCD07375466