N-Boc-4-carboxymethoxypiperidine 95%
Catalog # H32102.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
Specifications
About this item
CAS Number:161948-70-5
:
Formula:C₁₂H₂₁NO₅
:259.30 g/mol
Melting Pt:105…107 °C
:GHS07
Storage Temperature:Ambient
MDL Number:MFCD02178987