Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-Boc-4-oxo-L-proline-tert-butyl ester
N-Boc-4-oxo-L-proline-tert-butyl ester 97%
N-Boc-4-oxo-L-proline-tert-butyl ester 97%
N-Boc-4-oxo-L-proline-tert-butyl ester 97%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
N-Boc-4-oxo-L-proline-tert-butyl ester 97%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
166410-05-5
:
Formula:
C₁₄H₂₃NO₅
MDL Number:
MFCD06797013
:
285.35 g/mol
Melting Pt:
64…67 °C
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAH27375
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation