(S)-(-)-3,3'-Di-tert-butyl-5,5',6,6'-tetramethylbiphenyl-2,2'-diol 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:205927-03-3
:
Formula:C₂₄H₃₄O₂
MDL Number:MFCD01862439
:354.526 g/mol
Melting Pt:171…173 °C
Storage Temperature:Ambient