Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
tert-Butyl-3-oxopyrrolidine-1-carboxylate
tert-Butyl-3-oxopyrrolidine-1-carboxylate 97%
tert-Butyl-3-oxopyrrolidine-1-carboxylate 97%
tert-Butyl-3-oxopyrrolidine-1-carboxylate 97%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
tert-Butyl-3-oxopyrrolidine-1-carboxylate 97%
Supplier:
Thermo Fisher Scientific
About this item
CAS Number:
101385-93-7
:
Formula:
C₉H₁₅NO₃
MDL Number:
MFCD01631194
:
185.22 g/mol
Melting Pt:
34…38 °C
:
GHS07
Related Information
Documentation
SDS
msds_ALFAH27023
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation