Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-(Trimethylsilyl)diphenylacetylene
4-(Trimethylsilyl)diphenylacetylene 99%
4-(Trimethylsilyl)diphenylacetylene 99%
4-(Trimethylsilyl)diphenylacetylene 99%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
4-(Trimethylsilyl)diphenylacetylene 99%
Supplier:
Thermo Fisher Scientific
About this item
CAS Number:
136459-72-8
:
Formula:
C₁₇H₁₈Si
MDL Number:
MFCD09953470
:
250.42 g/mol
Melting Pt:
37…39 °C
Storage Temperature:
Ambient
Related Information
Documentation
SDS
msds_ALFAH26300
Videos
Thermo Scientific Acroseal 25 Years
Documentation