
2-Benzyloxy-2-methylpropan-1-ol 95%
Catalog # H26006.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
CAS Number:91968-71-7
:
Formula:C₁₁H₁₆O₂
MDL Number:MFCD08459314
:180.25 g/mol
Boiling Pt:80…85 °C (0.1 mmHg)
:GHS07
Storage Temperature:Ambient