
Imidazole-4-carboxaldehyde 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3034-50-2
:
Formula:C₄H₄N₂O
MDL Number:MFCD00173726
:96.09 g/mol
Melting Pt:174…177 °C
EINECS:221-227-3
:GHS07
Storage Temperature:Ambient