Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethyl-5-nitro-2-furoate
Ethyl-5-nitro-2-furoate 97%
Ethyl-5-nitro-2-furoate 97%
Ethyl-5-nitro-2-furoate 97%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Ethyl-5-nitro-2-furoate 97%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
943-37-3
:
Formula:
C₇H₇NO₅
MDL Number:
MFCD00092566
:
185.14 g/mol
Melting Pt:
98…101 °C
Storage Temperature:
Ambient
EINECS:
213-400-7
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAB24861
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation