Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl-4-hydroxy-2-hexynoate
Methyl-4-hydroxy-2-hexynoate 97%
Methyl-4-hydroxy-2-hexynoate 97%
Irritant
Methyl-4-hydroxy-2-hexynoate 97%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
Methyl-4-hydroxy-2-hexynoate 97%
Supplier:
Thermo Fisher Scientific
About this item
CAS Number:
112780-04-8
:
Formula:
C₇H₁₀O₃
MDL Number:
MFCD04039770
:
142.15 g/mol
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
SDS
msds_ALFAB24690
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation